Anti-VAMP7 Mouse Monoclonal Antibody [clone: 549115]

Supplier: R & D Systems
MAB6117
220021-340EA 594.37 USD
220021-340
Anti-VAMP7 Mouse Monoclonal Antibody [clone: 549115]
Antibodies
Type: Primary
Antigen: VAMP7
Clonality: Monoclonal
Clone: 549115
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR