Anti-TCF7L1 Mouse Monoclonal Antibody [clone: 622782]

Supplier: R & D Systems
MAB61161
102886-242EA 561.11 USD
102886-242
Anti-TCF7L1 Mouse Monoclonal Antibody [clone: 622782]
Antibodies
Type: Primary
Antigen: TCF7L1
Clonality: Monoclonal
Clone: 622782
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR