Anti-TCF7L1 Mouse Monoclonal Antibody [clone: 622722]

Supplier: R & D Systems
MAB6116
102886-240EA 561.11 USD
102886-240
Anti-TCF7L1 Mouse Monoclonal Antibody [clone: 622722]
Antibodies
Type: Primary
Antigen: TCF7L1
Clonality: Monoclonal
Clone: 622722
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR