Anti-IRF6 Mouse Monoclonal Antibody [clone: 503404]

Supplier: R & D Systems
MAB6114
220021-339EA 561.11 USD
220021-339
Anti-IRF6 Mouse Monoclonal Antibody [clone: 503404]
Antibodies
Type: Primary
Antigen: IRF6
Clonality: Monoclonal
Clone: 503404
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR