Anti-RORC Mouse Monoclonal Antibody [clone: 695935]

Supplier: R & D Systems
MAB61091
102886-236EA 577.02 USD
102886-236
Anti-RORC Mouse Monoclonal Antibody [clone: 695935]
Antibodies
Type: Primary
Antigen: RORC
Clonality: Monoclonal
Clone: 695935
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR