Anti-RORC Mouse Monoclonal Antibody [clone: 600214]

Supplier: R & D Systems
MAB6109
220021-337EA 647.88 USD
220021-337
Anti-RORC Mouse Monoclonal Antibody [clone: 600214]
Antibodies
Type: Primary
Antigen: RORC
Clonality: Monoclonal
Clone: 600214
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR