Anti-ADM Mouse Monoclonal Antibody [clone: 357923]

Supplier: R & D Systems
MAB6108
102886-234EA 577.02 USD
102886-234
Anti-ADM Mouse Monoclonal Antibody [clone: 357923]
Antibodies
Type: Primary
Antigen: ADM
Clonality: Monoclonal
Clone: 357923
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR