Anti-PPP1CA Mouse Monoclonal Antibody [clone: 490314]

Supplier: R & D Systems
MAB6105
220021-336EA 594.37 USD
220021-336
Anti-PPP1CA Mouse Monoclonal Antibody [clone: 490314]
Antibodies
Type: Primary
Antigen: PPP1CA
Clonality: Monoclonal
Clone: 490314
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR