Anti-PIK3R4 Mouse Monoclonal Antibody [clone: 576514]

Supplier: R & D Systems
MAB6104
220021-335EA 584.25 USD
220021-335
Anti-PIK3R4 Mouse Monoclonal Antibody [clone: 576514]
Antibodies
Type: Primary
Antigen: PIK3R4
Clonality: Monoclonal
Clone: 576514
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR