Anti-PKN1 Mouse Monoclonal Antibody [clone: 540421]

Supplier: R & D Systems
MAB6100
220021-334EA 577.02 USD
220021-334
Anti-PKN1 Mouse Monoclonal Antibody [clone: 540421]
Antibodies
Type: Primary
Antigen: PKN1
Clonality: Monoclonal
Clone: 540421
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR