Anti-SPRY1 Mouse Monoclonal Antibody [clone: 669534]

Supplier: R & D Systems
MAB6097
102886-232EA 577.02 USD
102886-232
Anti-SPRY1 Mouse Monoclonal Antibody [clone: 669534]
Antibodies
Type: Primary
Antigen: SPRY1
Clonality: Monoclonal
Clone: 669534
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR