Anti-CTTN Mouse Monoclonal Antibody [clone: 771716]

Supplier: R & D Systems
MAB6096
102886-230EA 561.11 USD
102886-230
Anti-CTTN Mouse Monoclonal Antibody [clone: 771716]
Antibodies
Type: Primary
Antigen: CTTN
Clonality: Monoclonal
Clone: 771716
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR