Anti-MAP3K3 Mouse Monoclonal Antibody [clone: 576240]

Supplier: R & D Systems
MAB6095
220021-332EA 594.37 USD
220021-332
Anti-MAP3K3 Mouse Monoclonal Antibody [clone: 576240]
Antibodies
Type: Primary
Antigen: MAP3K3
Clonality: Monoclonal
Clone: 576240
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR