Anti-RIT2 Mouse Monoclonal Antibody [clone: 456313]

Supplier: R & D Systems
MAB6094
220021-331EA 561.11 USD
220021-331
Anti-RIT2 Mouse Monoclonal Antibody [clone: 456313]
Antibodies
Type: Primary
Antigen: RIT2
Clonality: Monoclonal
Clone: 456313
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR