Anti-JARID2 Mouse Monoclonal Antibody [clone: 674702]

Supplier: R & D Systems
MAB6090
102886-228EA 577.02 USD
102886-228
Anti-JARID2 Mouse Monoclonal Antibody [clone: 674702]
Antibodies
Type: Primary
Antigen: JARID2
Clonality: Monoclonal
Clone: 674702
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR