Anti-HS3ST4 Mouse Monoclonal Antibody [clone: 712011]

Supplier: R & D Systems
MAB60851
102886-224EA 561.11 USD
102886-224
Anti-HS3ST4 Mouse Monoclonal Antibody [clone: 712011]
Antibodies
Type: Primary
Antigen: HS3ST4
Clonality: Monoclonal
Clone: 712011
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR