Anti-VIP Mouse Monoclonal Antibody [clone: 576721]

Supplier: R & D Systems
MAB6079
220021-326EA 594.37 USD
220021-326
Anti-VIP Mouse Monoclonal Antibody [clone: 576721]
Antibodies
Type: Primary
Antigen: VIP
Clonality: Monoclonal
Clone: 576721
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR