Anti-AVP Mouse Monoclonal Antibody [clone: 579021]

Supplier: R & D Systems
MAB6077
220021-324EA 633.42 USD
220021-324
Anti-AVP Mouse Monoclonal Antibody [clone: 579021]
Antibodies
Type: Primary
Antigen: AVP
Clonality: Monoclonal
Clone: 579021
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR