Anti-EPHA6 Rat Monoclonal Antibody [clone: 81605]

Supplier: R & D Systems
MAB6071
220021-323EA 763.57 USD
220021-323
Anti-EPHA6 Rat Monoclonal Antibody [clone: 81605]
Antibodies
Blockade of Receptor-ligand Interaction

Type: Primary
Antigen: EPHA6
Clonality: Monoclonal
Clone: 81605
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG1
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR