Anti-EPHA6 Rat Monoclonal Antibody [clone: 81610]

Supplier: R & D Systems
MAB607
220021-322EA 763.57 USD
220021-322
Anti-EPHA6 Rat Monoclonal Antibody [clone: 81610]
Antibodies
Type: Primary
Antigen: EPHA6
Clonality: Monoclonal
Clone: 81610
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR