Anti-CMV UL147/vCXC2 Mouse Monoclonal Antibody [clone: 146326]

Supplier: R & D Systems
MAB606
220021-321EA 763.57 USD
220021-321
Anti-CMV UL147/vCXC2 Mouse Monoclonal Antibody [clone: 146326]
Antibodies
Type: Primary
Antigen: CMV UL147/vCXC2
Clonality: Monoclonal
Clone: 146326
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR