Anti-MADCAM1 Mouse Monoclonal Antibody [clone: 683715]

Supplier: R & D Systems
MAB6056
102886-220EA 594.37 USD
102886-220
Anti-MADCAM1 Mouse Monoclonal Antibody [clone: 683715]
Antibodies
Type: Primary
Antigen: MADCAM1
Clonality: Monoclonal
Clone: 683715
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR