Anti-RCOR1 Mouse Monoclonal Antibody [clone: 658526]

Supplier: R & D Systems
MAB6047
102886-214EA 584.25 USD
102886-214
Anti-RCOR1 Mouse Monoclonal Antibody [clone: 658526]
Antibodies
Type: Primary
Antigen: RCOR1
Clonality: Monoclonal
Clone: 658526
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR