Anti-EDIL3 Mouse Monoclonal Antibody [clone: 670421]

Supplier: R & D Systems
MAB6046
102886-212EA 594.37 USD
102886-212
Anti-EDIL3 Mouse Monoclonal Antibody [clone: 670421]
Antibodies
Type: Primary
Antigen: EDIL3
Clonality: Monoclonal
Clone: 670421
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR