Anti-CCL5 Mouse Monoclonal Antibody [clone: 676117]

Supplier: R & D Systems
MAB6045
102886-200EA 561.11 USD
102886-200
Anti-CCL5 Mouse Monoclonal Antibody [clone: 676117]
Antibodies
Type: Primary
Antigen: CCL5
Clonality: Monoclonal
Clone: 676117
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Dog
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR