Anti-CSF3R Rat Monoclonal Antibody [clone: 723806]

Supplier: R & D Systems
MAB60391
102886-198EA 600.15 USD
102886-198
Anti-CSF3R Rat Monoclonal Antibody [clone: 723806]
Antibodies
Type: Primary
Antigen: CSF3R
Clonality: Monoclonal
Clone: 723806
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR