Anti-CSF3R Rat Monoclonal Antibody [clone: 680206]

Supplier: R & D Systems
MAB6039
102886-196EA 779.48 USD
102886-196
Anti-CSF3R Rat Monoclonal Antibody [clone: 680206]
Antibodies
Type: Primary
Antigen: CSF3R
Clonality: Monoclonal
Clone: 680206
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR