Anti-BMP10 Rat Monoclonal Antibody [clone: 654319]

Supplier: R & D Systems
MAB6038
102886-194EA 594.37 USD
102886-194
Anti-BMP10 Rat Monoclonal Antibody [clone: 654319]
Antibodies
Type: Primary
Antigen: BMP10
Clonality: Monoclonal
Clone: 654319
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR