Anti-BoNT-D Mouse Monoclonal Antibody [clone: 695004]

Supplier: R & D Systems
MAB6037
102886-192EA 594.37 USD
102886-192
Anti-BoNT-D Mouse Monoclonal Antibody [clone: 695004]
Antibodies
Type: Primary
Antigen Symbol:
Antigen Name: BoNT-D Light Chain
Antigen Synonyms:
Clonality: Monoclonal
Clone: 695004
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: C. botulinum
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR