Anti-CPB2 Mouse Monoclonal Antibody [clone: 650801]

Supplier: R & D Systems
MAB6036
102886-190EA 566.89 USD
102886-190
Anti-CPB2 Mouse Monoclonal Antibody [clone: 650801]
Antibodies
Type: Primary
Antigen: CPB2
Clonality: Monoclonal
Clone: 650801
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR