Anti-FBXO15 Mouse Monoclonal Antibody [clone: 612834]

Supplier: R & D Systems
MAB6035
102886-188EA 577.02 USD
102886-188
Anti-FBXO15 Mouse Monoclonal Antibody [clone: 612834]
Antibodies
Type: Primary
Antigen Symbol: FBXO15
Antigen Name: F-box protein 15
Antigen Synonyms: FBX15
Clonality: Monoclonal
Clone: 612834
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR