Anti-TTK Mouse Monoclonal Antibody [clone: 689913]

Supplier: R & D Systems
MAB6028
102886-184EA 577.02 USD
102886-184
Anti-TTK Mouse Monoclonal Antibody [clone: 689913]
Antibodies
Type: Primary
Antigen: TTK
Clonality: Monoclonal
Clone: 689913
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR