Anti-NME1 Mouse Monoclonal Antibody [clone: 642825]

Supplier: R & D Systems
MAB6019
220021-316EA 577.02 USD
220021-316
Anti-NME1 Mouse Monoclonal Antibody [clone: 642825]
Antibodies
Type: Primary
Antigen: NME1
Clonality: Monoclonal
Clone: 642825
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR