Anti-PCSK2 Mouse Monoclonal Antibody [clone: 694009]

Supplier: R & D Systems
MAB6018
102886-182EA 577.02 USD
102886-182
Anti-PCSK2 Mouse Monoclonal Antibody [clone: 694009]
Antibodies
Type: Primary
Antigen: PCSK2
Clonality: Monoclonal
Clone: 694009
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR