Anti-PDCD6 Mouse Monoclonal Antibody [clone: 722232]

Supplier: R & D Systems
MAB6015
102886-180EA 577.02 USD
102886-180
Anti-PDCD6 Mouse Monoclonal Antibody [clone: 722232]
Antibodies
Type: Primary
Antigen: PDCD6
Clonality: Monoclonal
Clone: 722232
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR