Anti-HSPA2 Mouse Monoclonal Antibody [clone: 520608]

Supplier: R & D Systems
MAB6010
220021-315EA 577.02 USD
220021-315
Anti-HSPA2 Mouse Monoclonal Antibody [clone: 520608]
Antibodies
Type: Primary
Antigen: HSPA2
Clonality: Monoclonal
Clone: 520608
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR