Anti-UCHL3 Mouse Monoclonal Antibody [clone: 677710]

Supplier: R & D Systems
MAB6008
102886-178EA 594.37 USD
102886-178
Anti-UCHL3 Mouse Monoclonal Antibody [clone: 677710]
Antibodies
Type: Primary
Antigen: UCHL3
Clonality: Monoclonal
Clone: 677710
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR