Anti-UCHL1 Mouse Monoclonal Antibody [clone: 671108]

Supplier: R & D Systems
MAB6007
102886-176EA 639.2 USD
102886-176
Anti-UCHL1 Mouse Monoclonal Antibody [clone: 671108]
Antibodies
Simple Western

Type: Primary
Antigen: UCHL1
Clonality: Monoclonal
Clone: 671108
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR