Anti-PARP1 Rat Monoclonal Antibody [clone: 53015]

Supplier: R & D Systems
MAB600
220021-313EA 763.57 USD
220021-313
Anti-PARP1 Rat Monoclonal Antibody [clone: 53015]
Antibodies
Type: Primary
Antigen: PARP1
Clonality: Monoclonal
Clone: 53015
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR