Anti-CBL Mouse Monoclonal Antibody [clone: 648005]

Supplier: R & D Systems
MAB5998
102886-174EA 584.25 USD
102886-174
Anti-CBL Mouse Monoclonal Antibody [clone: 648005]
Antibodies
Type: Primary
Antigen: CBL
Clonality: Monoclonal
Clone: 648005
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR