Anti-C3AR1 Mouse Monoclonal Antibody [clone: 534625]

Supplier: R & D Systems
MAB5995
220021-311EA 594.37 USD
220021-311
Anti-C3AR1 Mouse Monoclonal Antibody [clone: 534625]
Antibodies
Type: Primary
Antigen: C3AR1
Clonality: Monoclonal
Clone: 534625
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR