Anti-NRP1 Rat Monoclonal Antibody [clone: 761704]

Supplier: R & D Systems
MAB59941
102886-172EA 655.11 USD
102886-172
Anti-NRP1 Rat Monoclonal Antibody [clone: 761704]
Antibodies
Type: Primary
Antigen: NRP1
Clonality: Monoclonal
Clone: 761704
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR