Anti-LIFR Rat Monoclonal Antibody [clone: 673602]

Supplier: R & D Systems
MAB5990
102886-168EA 594.37 USD
102886-168
Anti-LIFR Rat Monoclonal Antibody [clone: 673602]
Antibodies
Type: Primary
Antigen: LIFR
Clonality: Monoclonal
Clone: 673602
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG1
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR