Anti-CFHR4 Mouse Monoclonal Antibody [clone: 640212]

Supplier: R & D Systems
MAB5980
102886-164EA 594.37 USD
102886-164
Anti-CFHR4 Mouse Monoclonal Antibody [clone: 640212]
Antibodies
Type: Primary
Antigen: CFHR4
Clonality: Monoclonal
Clone: 640212
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR