Anti-ENPP5 Mouse Monoclonal Antibody [clone: 500915]

Supplier: R & D Systems
MAB5979
220021-309EA 561.11 USD
220021-309
Anti-ENPP5 Mouse Monoclonal Antibody [clone: 500915]
Antibodies
Type: Primary
Antigen: ENPP5
Clonality: Monoclonal
Clone: 500915
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR