Anti-CRTC1 Rat Monoclonal Antibody [clone: 505531]

Supplier: R & D Systems
MAB5978
220021-308EA 561.11 USD
220021-308
Anti-CRTC1 Rat Monoclonal Antibody [clone: 505531]
Antibodies
Type: Primary
Antigen: CRTC1
Clonality: Monoclonal
Clone: 505531
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR