Anti-NUP85 Mouse Monoclonal Antibody [clone: 783403]

Supplier: R & D Systems
MAB5976
102886-162EA 561.11 USD
102886-162
Anti-NUP85 Mouse Monoclonal Antibody [clone: 783403]
Antibodies
Type: Primary
Antigen: NUP85
Clonality: Monoclonal
Clone: 783403
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR