Anti-IFNZ Rat Monoclonal Antibody [clone: 183707]

Supplier: R & D Systems
MAB5971
220021-306EA 763.57 USD
220021-306
Anti-IFNZ Rat Monoclonal Antibody [clone: 183707]
Antibodies
Type: Primary
Antigen: IFNZ
Clonality: Monoclonal
Clone: 183707
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR