Anti-IGFL3 Mouse Monoclonal Antibody [clone: 536825]

Supplier: R & D Systems
MAB5970
220021-305EA 561.11 USD
220021-305
Anti-IGFL3 Mouse Monoclonal Antibody [clone: 536825]
Antibodies
Type: Primary
Antigen: IGFL3
Clonality: Monoclonal
Clone: 536825
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR