Anti-GBA3 Mouse Monoclonal Antibody [clone: 728714]

Supplier: R & D Systems
MAB5969
102886-158EA 577.02 USD
102886-158
Anti-GBA3 Mouse Monoclonal Antibody [clone: 728714]
Antibodies
Type: Primary
Antigen Symbol: GBA3
Antigen Name: glucosidase, beta, acid 3 (gene/pseudogene)
Antigen Synonyms: CBG, CBGL1, GLUC, KLRP
Clonality: Monoclonal
Clone: 728714
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR